Learn More
Hach Company Clamp, Extension Thermometer
Supplier:  Hach Company 2656200
Encompass
Replacement clamp for use with the Cold Vapor Mercury Test. Hold thermometers and other glass apparatus firmly with this lightweight, safety-adjust clamp. Holds 6.4–12.7mm (1/4–1/2") diameter glass tubing/thermometers up to 178mm (7") from support rod. Tightening wing nut applies tension to the nickel-plated phosphor bronze jaws. Overall length: 210mm (81/4"). If attaching to rods, a 90° clamp holder is required. 1/pk
Catalog No. 50-199-6448