Learn More
Brady™ Clamp-on EZ Panel Loc System Circuit Breaker Device
Catalog No. 19103956 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-956 Red
1 options
Catalog No. 19-103-956 Supplier Brady™ Supplier No. 51254
  • $48.65
    $37.65 / Each of 1
    Save $11.00
    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Use on beakers that do not have a hole in the switch tongue. The Brady™; Clamp-on Ex Panel Loc System Circuit Breaker Device is easy to set up and ensures additional security for your circuit breakers.
  • For use on breakers that do not have a hole in the switch tongue, including multiple breakers with tie-bars
  • Turn thumbscrew to clamp lockout securely onto switch, then snap the cover closed to prevent thumbwheel from being loosened
Specifications
Color Red
Material Fiberglass-reinforced Polyurethane
Dimensions (L x W x H) 0.74 x 4 x 0.8 in.
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.