Learn More
Chemglass Life Sciences Flexible Clamp Kit, 12" Arm
SDP
Catalog No. 50871970
Encompass_Preferred
Flexible Clamp Kit, 12" Arm. Flexible clamp for use in clamping laboratory apparatus that are normally difficult to hold due to their size, shape or odd angles. Unique flexible arm can be placed in virtually any position or angle. Available with a choice of a 12 or 18 inch long flexible arm. Complete item consists of a two-prong clamp head, three-prong clamp head, spring clamp head, flexible arm and a lab frame connector with utility wrench. Base plate and bench clamps are available as optional items.
Catalog No. 50-871-970 Supplier Chemglass Life Sciences Supplier No. CG921401
Flexible Clamp Kit, 12" Arm. Flexible clamp for use in clamping laboratory apparatus that are normally difficult to hold due to their size, shape or odd angles. Unique flexible arm can be placed in virtually any position or angle. Available with a choice of a 12 or 18 inch long flexible arm. Complete item consists of a two-prong clamp head, three-prong clamp head, spring clamp head, flexible arm and a lab frame connector with utility wrench. Base plate and bench clamps are available as optional items.