Learn More
Research Products International Corp Clamp for Magnetic Platform, 250ml
SDP
Catalog No. 501978319
Encompass
MAGic Clamp Flask Clamps are for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators.
Magnetic flask clamp secures Erlenmeyer flask to a shaker platform for mixing or aerating operations.
Catalog No. 50-197-8319 Supplier Research Products International Corp Supplier No. 248575
MAGic Clamp Flask Clamps are for use with Incu-Shaker Mini and Incu-Shaker 10L model Shaking Incubators.
Magnetic flask clamp secures Erlenmeyer flask to a shaker platform for mixing or aerating operations.