Learn More
Kuhner Shaker Inc U-3000 FERNBACH CLAMP ET AL
Catalog No. NC0818975
Encompass
Clamp U-3000F - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 3000ml Fernbach or 5000mL Thomson flasks
Catalog No. NC0818975 Supplier Kuhner Shaker Inc Supplier No. SM313000F
Clamp U-3000F - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 3000ml Fernbach or 5000mL Thomson flasks