Learn More
Chemglass Life Sciences Heavy Duty Clamp Holder, Fits 1/4" to 5/8" Support Rod, White Epoxy Coated
SDP
Catalog No. 50143620
Encompass_Preferred
Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance. Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjustment and secure clamping power. Useful wherever clamping at 90¿ is required.
Catalog No. 50-143-620 Supplier Chemglass Life Sciences Supplier No. CG2040C01
Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance. Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjustment and secure clamping power. Useful wherever clamping at 90¿ is required.