Learn More
Wilmad Labglass CLAMP HLDR 1/4 STEEL THMBSCRW
Catalog No. 50802247
Encompass
Clamp Holder, Cast-Iron, w/ 1/4 in. Steel Thumbscrew. For use in lattice systems. Fits rods to 3/4 in. diameter
Catalog No. 50-802-247 Supplier Wilmad Labglass Supplier No. LG4622100
Clamp Holder, Cast-Iron, w/ 1/4 in. Steel Thumbscrew. For use in lattice systems. Fits rods to 3/4 in. diameter