Learn More
Cole-Parmer Cole-Parmer Jumbo Clamp Holder, Stainless Steel, Accommodates 0 to 0.83" Dia Rod s
Catalog No. NC1153642 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for holding clamps or ring stands to lab-frames, at a 90° angle.
Catalog No. NC1153642 Supplier Cole-Parmer Supplier No. 0802162
Ideal for holding clamps or ring stands to lab-frames, at a 90° angle.