Learn More
Chemglass Life Sciences Clamp Holder, Heavy Duty, Large
SDP
Catalog No. CG925201
Encompass_Preferred
Clamp Holder, Heavy Duty, Large. Heavy duty holder features greater grip strength and the ability to hold flat panels or shelves up to 3/4” (19mm) thick. Large thumb screws allow easy adjusting. Maximum grip size is 3/4” (19mm).
Catalog No. CG925201 Supplier Chemglass Life Sciences Supplier No. CG925201
Clamp Holder, Heavy Duty, Large. Heavy duty holder features greater grip strength and the ability to hold flat panels or shelves up to 3/4” (19mm) thick. Large thumb screws allow easy adjusting. Maximum grip size is 3/4” (19mm).