Learn More
Hach Company Clamp Holder
Supplier:  Hach Company 32600
Encompass
For rods or pipes between 1.1 and 2.2 cm (0.43 to 0.87") diameter in any type of right-angle clamp setup (horizontal or vertical). Comes with two 45 angle ribbed thumb screws. Not recommended for use with support ring stands.
Catalog No. 50-199-4250