Learn More
Chemglass Life Sciences Clamp Holder, Maximum rod grip size is 3/4 (19mm)
SDP
Catalog No. CG925001
Encompass_Preferred
Clamp Holder. Nickel-plated zinc alloy clamp holder has high tensile strength and corrosion resistance. Useful wherever clamping at 90 is required. Large thumb screws allow easy adjusting. Maximum rod grip size is 3/4 (19mm).
Catalog No. CG925001 Supplier Chemglass Life Sciences Supplier No. CG925001
Clamp Holder. Nickel-plated zinc alloy clamp holder has high tensile strength and corrosion resistance. Useful wherever clamping at 90 is required. Large thumb screws allow easy adjusting. Maximum rod grip size is 3/4 (19mm).