Learn More
Chemglass Life Sciences Clamp Holder, Premium
SDP
Catalog No. NC1115520
Encompass_Preferred
Clamp Holder, Premium, Nickel-plated zinc alloy clamp holder is ideal for holding clamps to lab frames. Use wherever clamping at 90 is required. Maximum rod grip size is 18mm.
Catalog No. NC1115520 Supplier Chemglass Life Sciences Supplier No. CG9250P01
Clamp Holder, Premium, Nickel-plated zinc alloy clamp holder is ideal for holding clamps to lab frames. Use wherever clamping at 90 is required. Maximum rod grip size is 18mm.