Learn More
Chemglass Life Sciences Clamp Holder, Swivel
SDP
Catalog No. 50871983
Encompass_Preferred
Clamp Holder, Swivel. Two clamp holders with a center swivel to allow tilting of clamp at any angle in parallel planes. Outside adjustment screw permits close proximity between items being held. Maximum grip size is 3/4” (19mm).
Catalog No. 50-871-983 Supplier Chemglass Life Sciences Supplier No. CG925501
Clamp Holder, Swivel. Two clamp holders with a center swivel to allow tilting of clamp at any angle in parallel planes. Outside adjustment screw permits close proximity between items being held. Maximum grip size is 3/4” (19mm).