Learn More
Chemglass Life Sciences Clamp, Horseshoe Style, 5.5in Flange
SDP
Catalog No. 501531091
Encompass_Preferred
Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.
Catalog No. 50-153-1091 Supplier Chemglass Life Sciences Supplier No. CG13905
Horseshoe shaped clamp used to connect CG-138 flat o-ring flange. Design allows clamp to be installed or removed easily. Made of cast aluminum with chemically resistant green epoxy coating. Three adjustable tension locking screws assure even clamping pressure.