Learn More
Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
Supplier:  Cole-Parmer 0456124
Encompass_Preferred
Enhance the performance of your Cole-Parmer mixerCompatible with Cole-Parmer Compact Digital Lab Mixer (50801-00)Chemical-resistant epoxy construction for harsh applicationsHeavy-duty clamp, made of cast aluminum zinc alloy with chemical-resistant epoxy coating, holds rods up to 1-1/8" dia. Handles made of Stainless Steel.
Catalog No. NC1381080