Learn More
Chemglass Life Sciences Clamp, Keck, Green, Standard Taper, Fits Joint Sizes 10/13 & 10/29
SDP
Catalog No. 50871959
Encompass_Preferred
Clamp, Keck, Green, Standard Taper, Fits Joint Sizes 10/13 & 10/29. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. 50-871-959 Supplier Chemglass Life Sciences Supplier No. CG14501
Clamp, Keck, Green, Standard Taper, Fits Joint Sizes 10/13 & 10/29. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.