Learn More
Chemglass Life Sciences Clamp, Keck, Orange, Standard Taper, Fits Joint Sizes 34/28 & 34/45
SDP
Catalog No. 50871961
Encompass_Preferred
Clamp, Keck, Orange, Standard Taper, Fits Joint Sizes 34/28 & 34/45. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.
Catalog No. 50-871-961 Supplier Chemglass Life Sciences Supplier No. CG14507
Clamp, Keck, Orange, Standard Taper, Fits Joint Sizes 34/28 & 34/45. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10.