Learn More
Chemglass Life Sciences Clamp, Keck, Violet, Standard Taper, Fits Joint Sizes 12/18 & 12/30
SDP
Supplier:  Chemglass Life Sciences CG14502
Encompass_Preferred
Clamp, Keck, Dark Yellow, Standard Taper, Fits Joint Sizes 40/35 & 40/50. Made of Delrin, the clamp is non-corrosive and will not scratch glass. Can be used to 140 C. Color coded for easy identification. SOLD IN PACKAGES OF 10. Image shown is CG-145-01 green clamp.
Catalog No. 50-871-960