Learn More
Wilmad Labglass CLAMP KTTL 4IN
Catalog No. 50801024
Encompass
Clamp, Reaction Vessel, for Conical Flange. For Flange Size: 4 in. Designed for easy removal of top portion of clamp w/out disturbing the body of the reaction flask. Complete w/ an extension rod for clamping to a laboratory stand.
Catalog No. 50-801-024 Supplier Wilmad Labglass Supplier No. LG8070100
Clamp, Reaction Vessel, for Conical Flange. For Flange Size: 4 in. Designed for easy removal of top portion of clamp w/out disturbing the body of the reaction flask. Complete w/ an extension rod for clamping to a laboratory stand.