Learn More
Grainger Benchmark Scientific Dedicated Platform, Holds 4 Micro Plates, 1 ea
Catalog No. 502749514
Encompass_Preferred
This dedicated platform is designed to securely accommodate up to 4 standard microplates. It is engineered for reliable use with orbital and incubation shaking systems.
  • Holds 4 standard microplates
  • Compatible with orbital shakers and incubation shakers
Catalog No. 50-274-9514 Supplier Grainger Supplier No. 14N129
This dedicated platform is designed to securely accommodate up to 4 standard microplates. It is engineered for reliable use with orbital and incubation shaking systems.
  • Holds 4 standard microplates
  • Compatible with orbital shakers and incubation shakers