Learn More
Wilmad Labglass CLAMP MCCARTER 4 1/2IN
Catalog No. 50801631
Encompass
Clamp, McCarter. Clamp Size: 4- 1/2 in. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.
Catalog No. 50-801-631 Supplier Wilmad Labglass Supplier No. LG7316106
Clamp, McCarter. Clamp Size: 4- 1/2 in. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.