Learn More
Wilmad Labglass CLAMP MCCARTER 5 1/2IN
Catalog No. 50800476
Encompass
Clamp, McCarter. Clamp Size: 5- 1/2 in. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.
Catalog No. 50-800-476 Supplier Wilmad Labglass Supplier No. LG7316108
Clamp, McCarter. Clamp Size: 5- 1/2 in. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.