Learn More
Wilmad Labglass CLAMP MCCARTER 57MM
Supplier:  Wilmad Labglass LG7316102
Encompass
Clamp, McCarter. Clamp Size: 57mm. Stainless steel horseshoe-shaped clamp used to connect flat flanges together.
Catalog No. 50-800-655
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.