Learn More
Cole-Parmer Extech 380947 Mini clamp meter, true rms, 7/8" clamp
Supplier:  Cole-Parmer 2684012
Encompass
Measures resistance and DC/AC voltage and current. Features low-battery and overrange indications, zero adjustment for accurate measurement of DC current, overload protection, and hold function. Meter measures true rms and frequency. Recall minimum or maximum values at the push of a button.
Catalog No. 50-992-987