Learn More
Grainger REED Clamp Meter, 1000A, True RMS, CAT III 600V
Catalog No. 502761676
Encompass_Preferred
The 1000A True RMS AC/DC clamp meter is a durable instrument designed for industrial maintenance professionals performing preventative and scheduled maintenance on electro-mechanical equipment. It features a double-molded plastic housing, a 50,000-count backlit LCD display with an analog bargraph, and a built-in non-contact voltage (NCV) detector with an LED indicator.
  • Measures AC/DC current/voltage, resistance, capacitance, frequency, duty cycle and temperature
  • Features True RMS AC measurements for accurate readings
  • Includes min/max function, display hold, relative mode, diode test and continuity check
  • Provides safety rating of CAT III 600V
  • Includes test leads, thermocouple wire probe, thermocouple adapter, 9V battery and soft carrying case
  • Offers autoranging or manual range selection and auto-shut off after 15 minutes
Catalog No. 50-276-1676 Supplier Grainger Supplier No. 161A80
The 1000A True RMS AC/DC clamp meter is a durable instrument designed for industrial maintenance professionals performing preventative and scheduled maintenance on electro-mechanical equipment. It features a double-molded plastic housing, a 50,000-count backlit LCD display with an analog bargraph, and a built-in non-contact voltage (NCV) detector with an LED indicator.
  • Measures AC/DC current/voltage, resistance, capacitance, frequency, duty cycle and temperature
  • Features True RMS AC measurements for accurate readings
  • Includes min/max function, display hold, relative mode, diode test and continuity check
  • Provides safety rating of CAT III 600V
  • Includes test leads, thermocouple wire probe, thermocouple adapter, 9V battery and soft carrying case
  • Offers autoranging or manual range selection and auto-shut off after 15 minutes