Learn More
Chemglass Life Sciences Nylon Tubing Clamp, Clamping Range: .475-.536in, 12.07-13.61mm
SDP
Catalog No. 501311046
Encompass_Preferred
Nylon Tubing Clamp, Clamping Range: .475-.536in, 12.07-13.61mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release
Catalog No. 50-131-1046 Supplier Chemglass Life Sciences Supplier No. CG15213
Nylon Tubing Clamp, Clamping Range: .475-.536in, 12.07-13.61mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release