Learn More
Chemglass Life Sciences Nylon Tubing Clamp, Clamping Range: .840-.940in, 22.10-23.88mm
SDP
Catalog No. 501311051
Encompass_Preferred
Nylon Tubing Clamp, Clamping Range: .840-.940in, 22.10-23.88mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release
Catalog No. 50-131-1051 Supplier Chemglass Life Sciences Supplier No. CG15218
Nylon Tubing Clamp, Clamping Range: .840-.940in, 22.10-23.88mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release