Learn More
Grainger Grainger Clamp, Galvanized Steel, Vinyl Coated, 1/4 in. OD, 50/Pk.
Catalog No. 502794198
Encompass_Preferred
This clamp is designed for secure fastening of components with an outside diameter of up to 1/4 in. Constructed from hot-dipped galvanized steel, it provides reliable structural support and includes a vinyl coating to protect surfaces and enhance grip. The clamp is engineered for durability in various environments, offering effective corrosion resistance.
  • Fits a maximum outside diameter of 1/4 in.
  • Constructed with a 1/2 in. width for stability.
  • Features hot-dipped galvanized steel for long-term corrosion resistance.
  • Includes a vinyl coating for surface protection.
  • Operates within a temperature range of -30°F to +180°F.
  • Chem-treated G-90 per ASTM A653/A653M.
  • Supplied in a pack of 50 units.
Catalog No. 50-279-4198 Supplier Grainger Supplier No. 1AYZ4
This clamp is designed for secure fastening of components with an outside diameter of up to 1/4 in. Constructed from hot-dipped galvanized steel, it provides reliable structural support and includes a vinyl coating to protect surfaces and enhance grip. The clamp is engineered for durability in various environments, offering effective corrosion resistance.
  • Fits a maximum outside diameter of 1/4 in.
  • Constructed with a 1/2 in. width for stability.
  • Features hot-dipped galvanized steel for long-term corrosion resistance.
  • Includes a vinyl coating for surface protection.
  • Operates within a temperature range of -30°F to +180°F.
  • Chem-treated G-90 per ASTM A653/A653M.
  • Supplied in a pack of 50 units.