Learn More
Grainger Approved Vendor Cushion Clamp, Galvanized Steel, 5/16 in. Max OD, 50/Pk.
Catalog No. 502794240
Encompass_Preferred
These loop-style cushion clamps provide secure mounting for tubing and piping applications. The clamp is designed to slip over the pipe and pinch closed at the ends, where it is fastened to a flat surface with a screw. Constructed from galvanized steel with a vinyl plastisol cushion, these clamps offer reliable support and vibration dampening.
  • Galvanized steel construction
  • Vinyl plastisol cushion material
  • Maximum outside diameter capacity of 5/16 in.
  • Mounting hole diameter of 13/32 in.
  • Operating temperature range of -30°F to 180°F
  • Compliant with ASTM A240/A666, A653/A653M, and B209 standards
Catalog No. 50-279-4240 Supplier Grainger Supplier No. 1AYZ7
These loop-style cushion clamps provide secure mounting for tubing and piping applications. The clamp is designed to slip over the pipe and pinch closed at the ends, where it is fastened to a flat surface with a screw. Constructed from galvanized steel with a vinyl plastisol cushion, these clamps offer reliable support and vibration dampening.
  • Galvanized steel construction
  • Vinyl plastisol cushion material
  • Maximum outside diameter capacity of 5/16 in.
  • Mounting hole diameter of 13/32 in.
  • Operating temperature range of -30°F to 180°F
  • Compliant with ASTM A240/A666, A653/A653M, and B209 standards