Learn More
Electron Microscopy Sciences Big Eye™ Illuminated Magnilite Clamp-On, 2.0x
SDP
Catalog No. 5033202
Encompass_Preferred
"Big Eye" is one of our largest magnilites with a 7.5" optical quality lens. Wrapped in a ESD safe shade, it includes a 10" (250cm) shadowless 34-watt cirline bulb. 10 years mechanical warranty. 43" Ultraflex arms.
Magnification: 2.0x
Base: Clamp-on
Bulb: Cool white
Catalog No. 50-332-02 Supplier Electron Microscopy Sciences Supplier No. 6820210
"Big Eye" is one of our largest magnilites with a 7.5" optical quality lens. Wrapped in a ESD safe shade, it includes a 10" (250cm) shadowless 34-watt cirline bulb. 10 years mechanical warranty. 43" Ultraflex arms.
Magnification: 2.0x
Base: Clamp-on
Bulb: Cool white