Learn More
Brady™ Clamp-On Circuit Breaker Lockout
Catalog No. 17607635 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
6 Pieces
For Use With (Equipment):
120/277 V Circuit Breaker
480/600 V Circuit Breaker
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Equipment)
17-607-635 6 Pieces 480/600 V Circuit Breaker
17-607-633 6 Pieces 120/277 V Circuit Breaker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-607-635 Supplier Brady™ Supplier No. 176498
Add to Cart
Add to Cart
Experience reliable lockout safety and effortless installation with the Clamp-On Circuit Breaker Lockout. Simply twist the proprietary thumbwheel design for tool-free application.
Experience reliable lockout safety and effortless installation with the Clamp-On Circuit Breaker Lockout. Simply twist the proprietary thumbwheel design for tool-free application.
  • Fits a wide range of single-pole and internal-trip multi-pole breakers. Detachable cleat expands the range of applicable breakers.
  • Made from fiberglass-reinforced nylon for durability in demanding environments.
  • Tamper-resistant features help prevent unauthorized removal.
  • Convenient size for easy transport and storage.
  • Accepts padlock with shackle diameter up to 0.28 in.
Specifications
Product Type Circuit Breaker Lockout
Material Polycarbonate
Description Clamp-on Circuit Breaker
For Use With (Equipment) 480/600 V Circuit Breaker
Includes (6) Attachable Cleat for Circuit Breaker Lockout Device
Quantity 6 Pieces