Learn More
Cole-Parmer Extech 382357 Clamp-On Ground Resistance Tester
Catalog No. 50992594 Shop All Cole-Parmer Products
Encompass
Enables non-contact measurements of ground conductors without need for auxiliary ground spikes
FEATURES
  • Autoranging ground resistance measurements from 0.03 O to 1500 O with 0.02 O resolution
  • True RMS AC Leakage current range of 0.2mA to 15.00A
  • Programmable datalogging with 116 data points
  • Programmable Hi/Lo alarm
  • 0.9" (23mm) Jaw size for large ground rods
  • Simplifies ground resistance measurements on multiple point ground systems
  • Verification resistors confirm accuracy of meter
  • Automatic Self Calibration when powered on
  • 9999 Count LCD display with Data Hold
  • Electrical Noise Detection feature prevents inaccurate readings
  • CAT III-300V, CAT II-600V; CE
  • One year warranty
Catalog No. 50-992-594 Supplier Cole-Parmer Supplier No. 2000314
Enables non-contact measurements of ground conductors without need for auxiliary ground spikes
FEATURES
  • Autoranging ground resistance measurements from 0.03 O to 1500 O with 0.02 O resolution
  • True RMS AC Leakage current range of 0.2mA to 15.00A
  • Programmable datalogging with 116 data points
  • Programmable Hi/Lo alarm
  • 0.9" (23mm) Jaw size for large ground rods
  • Simplifies ground resistance measurements on multiple point ground systems
  • Verification resistors confirm accuracy of meter
  • Automatic Self Calibration when powered on
  • 9999 Count LCD display with Data Hold
  • Electrical Noise Detection feature prevents inaccurate readings
  • CAT III-300V, CAT II-600V; CE
  • One year warranty