Learn More
Wilmad Labglass CLAMP PINCH LOCKING SZ 35
Supplier:  Wilmad Labglass LG1045112
Encompass
Clamp, Pinch, Spherical Joints, Stainless Steel. Clamp Size: 35. This clamp is for use w/ spherical joints and is manufactured from stainless steel. Supplied w/ a screw type locking devise.
Catalog No. 50-802-160
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.