Learn More
Hach Company Clamp, Pinchcock, Day
Supplier:  Hach Company 68206
Encompass
Nickel-plated, steel wire can be used on tubing up to 10 mm (0.39") O.D. Length, 6.7 cm (2.6").
Catalog No. 50-199-4425