Learn More
Chemglass Life Sciences Clamp, PTFE, 14/20
SDP
Catalog No. 50871962
Encompass_Preferred
Clamp, PTFE, 14/20. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.
Catalog No. 50-871-962 Supplier Chemglass Life Sciences Supplier No. CG145T01
Clamp, PTFE, 14/20. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.