Learn More
Chemglass Life Sciences Clamp, PTFE, 19/22
SDP
Catalog No. 501531111
Encompass_Preferred
Clamp, PTFE, 19/22. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.
Catalog No. 50-153-1111 Supplier Chemglass Life Sciences Supplier No. CG145T02
Clamp, PTFE, 19/22. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.