Learn More
Chemglass Life Sciences Clamp, PTFE, 24/40
SDP
Catalog No. 50871964
Encompass_Preferred
Clamp, PTFE, 24/40. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.
Catalog No. 50-871-964 Supplier Chemglass Life Sciences Supplier No. CG145T03
Clamp, PTFE, 24/40. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.