Learn More
Chemglass Life Sciences Clamp, PTFE, 29/26-29/42
SDP
Catalog No. 501531112
Encompass_Preferred
Clamp, PTFE, 29/26-29/42. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.
Catalog No. 50-153-1112 Supplier Chemglass Life Sciences Supplier No. CG145T04
Clamp, PTFE, 29/26-29/42. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.