Learn More
Chemglass Life Sciences Clamp, PTFE, 34/45
SDP
Numéro de catalogue. 50871966
Encompass_Preferred
Clamp, PTFE, 34/45. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.
Numéro de catalogue. 50-871-966 Fournisseur Chemglass Life Sciences Code fournisseur CG145T05
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Clamp, PTFE, 34/45. PTFE molded clamps with a steel core can withstand temperatures up to 250°C and are totally chemically resistant and inert.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.