Learn More
Chemglass Life Sciences Clamp, PTFE, 45/50
SDP
Catalog No. 501531113
Encompass_Preferred
Clamp, PTFE, 45/50. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.
Catalog No. 50-153-1113 Supplier Chemglass Life Sciences Supplier No. CG145T06
Clamp, PTFE, 45/50. PTFE molded clamps with a steel core can withstand temperatures up to 250C and are totally chemically resistant and inert.