Learn More
Wilmad Labglass CLAMP QUICK RELEASE 150MM
Catalog No. 50801825
Encompass
Clamp, Quick Release, For Duran Flange. Fits Flange ID: 150mm. Quick release clamp made of stainless steel. Has screw adjustment for tension control to flange.
Catalog No. 50-801-825 Supplier Wilmad Labglass Supplier No. LG4632106
Clamp, Quick Release, For Duran Flange. Fits Flange ID: 150mm. Quick release clamp made of stainless steel. Has screw adjustment for tension control to flange.