Learn More
Chemglass Life Sciences Clamp, Stainless Steel, Quick Release, 100mm Duran Flange
SDP
Catalog No. NC0569863
Encompass_Preferred
Made entirely from stainless steel, the clamp has three sections that firmly hold reaction vessel and lid together. Quick release mechanism has a screw adjustment for tension applied to the flange. 100mm Schott Duran Flange
Catalog No. NC0569863 Supplier Chemglass Life Sciences Supplier No. CG14102
Made entirely from stainless steel, the clamp has three sections that firmly hold reaction vessel and lid together. Quick release mechanism has a screw adjustment for tension applied to the flange. 100mm Schott Duran Flange