Learn More
Chemglass Life Sciences Clamp, Reactor Support, 100mm
SDP
Catalog No. 50883304
Encompass_Preferred
Clamp, Reactor Support, 100mm. Similar to CG-1947, support clamp is designed to fit just below the flange of all Chemglass Reactors and permits Reactor lid to be detached without removing the Reactor body from the support clamp. Improved clamp is hinged on one side to swing open and allow complete access to install/remove reactor while clamp is still attached to lattice or benchtop support stand. Removable tightening knob holds Reactor secure. Clamps are constructed of stainless steel with a 1/2" OD X 9.5” long extension rod for connection to lattice and benchtop support stands. The inner portion of the clamp is lined with a high temperature red silicone rubber to prevent direct contact between the stainless steel and glass Reactor.PLEASE NOTE: IN ADDITION TO USING A CG-1947-A CLAMP, ALL REACTORS SHOULD BE SUPPORTED FROM BELOW USING THE CG-1948 SUPPORT RING.
Catalog No. 50-883-304 Supplier Chemglass Life Sciences Supplier No. CG1947A100
Clamp, Reactor Support, 100mm. Similar to CG-1947, support clamp is designed to fit just below the flange of all Chemglass Reactors and permits Reactor lid to be detached without removing the Reactor body from the support clamp. Improved clamp is hinged on one side to swing open and allow complete access to install/remove reactor while clamp is still attached to lattice or benchtop support stand. Removable tightening knob holds Reactor secure. Clamps are constructed of stainless steel with a 1/2" OD X 9.5” long extension rod for connection to lattice and benchtop support stands. The inner portion of the clamp is lined with a high temperature red silicone rubber to prevent direct contact between the stainless steel and glass Reactor.PLEASE NOTE: IN ADDITION TO USING A CG-1947-A CLAMP, ALL REACTORS SHOULD BE SUPPORTED FROM BELOW USING THE CG-1948 SUPPORT RING.