Learn More
Chemglass Life Sciences Clamp, Sanitary, 1", Heavy Duty, Single Pin, 304 Stainless Steel
SDP
Catalog No. 501311064
Encompass_Preferred
Clamp, Sanitary, 1", Heavy Duty, Single Pin, 304 Stainless Steel. Single pin style heavy duty sanitary clamp made from 304 stainless steel. For use with our sanitary adapters.
Catalog No. 50-131-1064 Supplier Chemglass Life Sciences Supplier No. CG152C10
Clamp, Sanitary, 1", Heavy Duty, Single Pin, 304 Stainless Steel. Single pin style heavy duty sanitary clamp made from 304 stainless steel. For use with our sanitary adapters.