Learn More
Chemglass Life Sciences Clamp, Sanitary, 3/4", Heavy Duty, Single Pin, 304 Stainless Steel
SDP
Catalog No. 501311063
Encompass_Preferred
Clamp, Sanitary, 3/4", Heavy Duty, Single Pin, 304 Stainless Steel. Single pin style heavy duty sanitary clamp made from 304 stainless steel. For use with our sanitary adapters.
Catalog No. 50-131-1063 Supplier Chemglass Life Sciences Supplier No. CG152C07
Clamp, Sanitary, 3/4", Heavy Duty, Single Pin, 304 Stainless Steel. Single pin style heavy duty sanitary clamp made from 304 stainless steel. For use with our sanitary adapters.