Learn More
Chemglass Life Sciences Clamp, Chain, Small
SDP
Catalog No. CG922001
Encompass_Preferred
Clamp, Chain, Small. Small size chain clamps to accommodate up to 6 1/2” (170mm) O.D. Quick slip-on chain connection has large, easy-to-turn adjusting knob. Extension arm has a length of 6” (150mm). Grip is vinyl coated.
Catalog No. CG922001 Supplier Chemglass Life Sciences Supplier No. CG922001
Clamp, Chain, Small. Small size chain clamps to accommodate up to 6 1/2” (170mm) O.D. Quick slip-on chain connection has large, easy-to-turn adjusting knob. Extension arm has a length of 6” (150mm). Grip is vinyl coated.