Learn More
Wilmad Labglass CLAMP ST METAL SZ 24
Catalog No. 50802253
Encompass
Joint Clamp, Metal, for Standard Taper joints, Fits Joint Sizes: 24/25, 24/40. Clamp is lacquer coated to avoid scratching glass. Joints are secure under moderate pressure to 200degreeC.
Catalog No. 50-802-253 Supplier Wilmad Labglass Supplier No. LG1006104
Joint Clamp, Metal, for Standard Taper joints, Fits Joint Sizes: 24/25, 24/40. Clamp is lacquer coated to avoid scratching glass. Joints are secure under moderate pressure to 200degreeC.