Learn More
Chemglass Life Sciences Clamp, Stainless Steel, Quick Release, 150mm Duran Flange
SDP
Catalog No. CG14104
Encompass_Preferred
Clamp, Stainless Steel, Quick Release, 150mm Duran Flange. Made entirely from stainless steel, the clamp has three sections that firmly hold reaction vessel and lid together. Quick release mechanism has a screw adjustment for tension applied to the flange. 150mm Duran Flange.
Catalog No. CG14104 Supplier Chemglass Life Sciences Supplier No. CG14104
Clamp, Stainless Steel, Quick Release, 150mm Duran Flange. Made entirely from stainless steel, the clamp has three sections that firmly hold reaction vessel and lid together. Quick release mechanism has a screw adjustment for tension applied to the flange. 150mm Duran Flange.