Learn More
Grainger Clamp Steel 1-1/4 In
Catalog No. 502752613
Encompass_Preferred
The Kindorf Angler Pipe Clamp is a steel pipe clamp designed for hanging pipes from ceilings or walls. It has a load capacity of 150 lbs and fits pipe sizes from 1/2" to 2". The clamp is made from high strength steel with a durable powder coat finish and features a unique angled design to provide stability. The clamp opens to fit over the pipe and tightens with a hex nut to firmly grip the pipe. Overall dimensions are 3-1/2" W x 4-1/2" H x 2-3/4" D. This heavy duty clamp securely mounts pipes in commercial, industrial, and residential settings.
Catalog No. 50-275-2613 Supplier Grainger Supplier No. 14L805
The Kindorf Angler Pipe Clamp is a steel pipe clamp designed for hanging pipes from ceilings or walls. It has a load capacity of 150 lbs and fits pipe sizes from 1/2" to 2". The clamp is made from high strength steel with a durable powder coat finish and features a unique angled design to provide stability. The clamp opens to fit over the pipe and tightens with a hex nut to firmly grip the pipe. Overall dimensions are 3-1/2" W x 4-1/2" H x 2-3/4" D. This heavy duty clamp securely mounts pipes in commercial, industrial, and residential settings.