Learn More
Hach Company Clamp, Two-Prong, Swivel
Supplier:  Hach Company 2503400
Encompass
Perfect for cylinder shaped glassware. Swivel feature allows for complete circular adjustments. Clamp complete with both slip-on vinyl and fiberglass grip sleeves. Length 5 - 7/16" (138 mm). Open diameter is 3" (77 mm).
Catalog No. 50-199-6101